missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rubbermaid™ Utility Carts
Rubbermaid Commercial Heavy-Duty Service Cart is perfect for transporting materials, supplies, and heavy loads in almost any environment.
Manufacturer: Rubbermaid™ 4525BLK
Description
- Ergonomically designed push handle provides optimum hand placement and push height improving control and worker safety
- Molded-in storage compartments, holsters, hooks, and notches
- Best-in-class casters to meet the most demanding environments
Specifications
33.3 in. | |
500 lb. | |
PP (Polypropylene) | |
113.4kg | |
84.6cm | |
111.4cm | |
43.86 x 25.86 x 33.3 in. (111.4 x 65.7 x 84.6cm) | |
Designed for transport of oversized, bulky items | |
Medium |
25.86 in. | |
226.8kg | |
250 lb. | |
Black | |
43.86 in. | |
65.7cm | |
2 | |
Ergonomically push handle with hand guard, molded-in customizable fittings, 2 flat shelf, rounded corners, large nonmarking casters, molded-in storage compartments, holsters, hooks and notches |