Rubbermaid™ HYGEN™ Dry Dusting Mops
Collects and holds dust and dirt mechanically and electrostatically
Manufacturer:  Rubbermaid™ Q41200WH
 View more versions of this product
Catalog No. 19-130-0863
Description
Description
  • High pile and looped fringe construction
  • Dense 3 - 5 micrometer quality fiber
  • Hook-and-loop backing for harsh laundering conditions
  • Double finished edges
  • Available with or without fringe
Specifications
Specifications
White
0.5 in.
5.5 in.
Dry Mop
For superior dusting results
18.5 in.
Microfiber