Eppendorf™ Shaker Flask Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Supplier:  Eppendorf™ M11919909DUP
Catalog No. 14-278-121
Description
Description
  • Dedicated platforms for specific-sized Erlenmeyer or Fernbach flasks
  • Supplied with clamps
Capacities listed are the most vessels of each size that will fit in a balanced pattern.