Eppendorf™ Gable Covers for Model C76 Water Bath Shaker
For C76 Water Bath Shaker
$1,370.64 - $1,370.64
Specifications
For Use With (Equipment) Model C76 water bath shaker
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
14-284-36 Eppendorf™
M12312000
Stainless Steel Each for $1,370.64
14-284-37 Eppendorf™
M12312010
Plexiglass™ Each for $1,370.64
Specifications
Description
  • Plexiglas™ cover permits temperatures up to 60°C Stainless-steel cover required for temperatures above 60°C
Description & Specifications
Specifications
Model C76 water bath shaker