missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neutec Proficlave PC10 and PC20 Media Preparator

Fully automated bench top agar/media preparator for fast processing of upto 16L of media in one hour

Specifications

Certifications/Compliance CE
Accuracy 0.1°C
Wattage 3500 w
View More Specs

Includes: Stainless steel cuvet, built-in pressure pump against flash boiling, extra-strong magnetic stirrer, sterile filter, thermal printer

Products 2
Catalog Number Mfr. No. Cycle Time Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Cycle Time Volume (Metric) Price Quantity  
14-910-317 View Documents Neutec Group Inc
9000610003
60 minutes 0.5 to 9 L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14-910-318 View Documents Neutec Group Inc
NTBT620003
85 minutes 0.5 to 16 L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Features enhanced user programming capabilities, operating parameter printout (integrated in front panel), external control functions (front panel connection), best stirring control and full metal housing. It accurately maintains the agar in any stage of the single or double cycle process at its temperature, even for hours!

  • Stores up to 20 user-defined programs (single or double cycle programs) for 3 different functional modes
  • Water jacket design enables even heating within the vessel
  • Atmospheric pressure measure and boiling point correction for reliable sterilization
  • Fully-controlled stirrer with variable speeds and enhanced magnetic coupling without seals or shafts
  • PT100 controlled temperature probe for agar and surrounding water jacket
  • Easy vessel access—no bolts or tools required
  • Multiple purpose (media sterilizer, bench top autoclave and stable water bath)

  • Multilingual software
  • Mechanical pressure gauge operates independently of power supply
  • Includes pressure sensor and valve with safety thermostat
  • Program data, parameters and operating status displayed via LED backlit graphic display
  • Timer function

PC10 Media Preparator

  • Prepares agar from 0.5 up to 9 liters
  • Cycle Speed: 60 min. from start to finish
  • D x W x H: 565 x 535 x 500mm
PC20 Media Preparator
  • Prepares agar from 0.5 up to 16 liters
  • Cycle Speed: 85 min. from start to finish
  • D x W x H: 565 x 535 x 580mm

Healthcare and diagnostic, Food research and quality assurance, Veterinary diagnostics, Agriculture research and quality assurance labs and Universities, Pharma and Biotech industries

Certifications

CE marked

Specifications

Specifications

CE
0.1°C
3500 w