missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neutec Group Poly Stainer

Programmable microbiology and histology staining instrument provides rapid, reproducible results

Manufacturer:  Neutec Group NT5301

Catalog No. 14-910-357

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: One slide basket, four plastic containers with lids

Description

Description

The Poly Stainer can automate just about any manual staining process. Some examples are: Gram stain, Acid Fast, Trichrome, Wright's stain, Reticulocyte stain, Bone Marrow staining, Gram Positive, Gram Negative

  • Unit is comprised of four staining chambers, a rinsing chamber, and a drying chamber
  • Stainless-steel container holder can be easily removed and cleaned
  • No risk to the electronic components due to their elevated position
  • Optional quick drain system empties the water bath to prevent overflow
  • Ten programs option each with 10 different positions
  • 20 slides capacity per basket

Order Info

For 115VAC 60Hz, 250W; L x W x H: 27.6 × 17.3 × 15.7 in. (70 × 44 × 40cm)

Specifications

Specifications

115VAC 60Hz
20
40cm
70cm
115VAC
One slide basket, four plastic containers with lids
Neutec™ Poly Stainer
15.7 in.
27.6 in.
17.3 in.
60Hz
SDS
Documents
Product Certifications
Promotions

Promotions