missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Neutec™ Petriswiss™ PS 20 Automated Petri Dish Filling System

Fills 20 dishes directly in the PetriRack™—no manual stacking required!

Manufacturer:  Neutec Group Inc BT800000

Catalog No. 14-910-319

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: PetriRack, feed rack, power cord (U.S.)

Description

Description

  • Compact, variable filling unit
  • Ideal for production of small to medium special-purpose agar batches
  • 20 agar plates are filled and stacked within 2 minutes
  • Intuitive software is accessible via touchscreen
  • Accommodates twenty 84 to 96mm dishes
  • Utilizes beveled edge/automation plates
  • Convenient PetriRack stores dishes for transport, refrigeration, incubation, or autoclaving

Specifications

Specifications

Petri Dish Filling System
40 w
20 kg
Petrirack, feed rack, power cord
44 lb.
Documents