Learn More
Neutec Group PetriSphere™ Anaerobic Gas Generating System
Generates atmospheres suitable for the culturing of anaerobic, microaerophilic and capnophilic bacteria in common anaerobic jars
Specifications
For Use With (Application) | Culture and recovery of oxygen-sensitive organisms |
---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
14-910-321
|
Neutec Group
NTBTIL.112011-04 |
PetriSphere Anaerobic Gas System |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
14-910-389
|
Neutec Group
NTBTIL.112011-05 |
Processing Unit |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The PetriSphere replaces the high cost of a traditional Glove Box, eliminates high consumable costs of sachet based anaerobic jars and reduces the handling costs of gas cylinders, supplies and waste products. The system consists of a microprocessor-based controller connected with a diaphragm vacuum pump and a nitrogen gas reservoir. Carefully controlled vacuum evacuation of the original gas and replacement with oxygen free gas produces critical improvements over traditional techniques. The container evacuates to the pressure indicated, vented with the reaction gas connected to the system, with the pre-programmed number of cycles processed automatically.
Innovative Design
- Extremely rapid oxygen depletion (anaerobic atmosphere)
- Flexible level of oxygen depletion (microaerophilic atmosphere)
- Creates multiple simultaneous atmospheres using multiple jars
- Convenient, affordable, with no additional consumables needed
- Increases lab productivity by applying automation
- Easily program vacuum, gas exchange pressure and the number of cycles needed
- Achieve your desired atmosphere by simply pressing the start button; process starts and runs fully automatically
- Special quick connect adaptors provide convenient, safe and time-saving operation
Specifications
Culture and recovery of oxygen-sensitive organisms |