Neutec Group Eddy Jet™ 2 Spiral Plating System
For spiral inoculation with disposable syringes
Manufacturer:  Neutec Group NT3700
Catalog No. 14-910-397
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes: 1000 syringes and 1000 sample beakers. Both irradiated with gamma rays.
Description
Description
Sterile gamma-irradiated disposable micro-syringe for each new sample eliminates cross contamination and ensures true sampling sterility
  • No use of bleach or requirement for a vacuum pump increases lab productivity
  • Plates are processed in well under 30 seconds
  • User–friendly programming software interface controlled from a color touch panel with intuitive icon operation
  • Ethernet, 2xUSB ports, and 2xRS-232 ports that allow connection to printer and barcode reader
  • Operation recordings can be directly dumped into a LIMS
  • Up to 22 different spiral distributions: the spiral plater with the widest range of spiral distributions
Specifications
Specifications
19.7 in.
Eddy Jet™ 2 Spiral Plating System
42 cm
20 kg
50 cm
16.5 in.
44 lb.
20.3 in.