missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Neurotrophin Receptor/NGF-Receptor (p75 NGFR) Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1, Mouse Monoclonal Antibody.

$277.55 - $1,509.45

Specifications

Antigen Neurotrophin Receptor/NGF-Receptor (p75superscriptNGFRsuperscript) Ab-1
Clone NGFR5
Applications Flow Cytometry
Classification Monoclonal
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS394B Thermo Scientific™ Lab Vision™
MS394B
200μg/mL Biotin 1mL Each for $1,509.45
MS394B0 Thermo Scientific™ Lab Vision™
MS394B0
200μg/mL Biotin 100μL Each for $371.61
MS394B1 Thermo Scientific™ Lab Vision™
MS394B1
200μg/mL Biotin 500μL Each for $1,010.84
MS394P Thermo Scientific™ Lab Vision™
MS394P
200μg/mL Unlabeled 1mL Each for $1,188.61
MS394P0 Thermo Scientific™ Lab Vision™
MS394P0
200μg/mL Unlabeled 100μL Each for $277.55
MS394P1 Thermo Scientific™ Lab Vision™
MS394P1
200μg/mL Unlabeled 500μL Each for $775.08
MS394P1ABX Thermo Scientific™ Lab Vision™
MS394P1ABX
1mg/mL Unlabeled 100μL Each for $854.10
MS394PABX Thermo Scientific™ Lab Vision™
MS394PABX
1mg/mL Unlabeled 200μL Each for $1,257.88
Specifications

Description

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1, Mouse Monoclonal Antibody.

NGF is important for the development, differentiation, and survival of variety of neuronal and non-neuronal cells. Its action is mediated by binding two distinct receptors, the high affinity p140 and low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR).

Host Species: Mouse

Clone: NGFR5

Isotype: IgG1/κ

Species Reactivity: Human, Monkey (Cebus apella, Macaca fascicularis, mulatta, and nemestrina), Baboon (Papio anubis and Cynocephalus), Cat, Rabbit, Ferret. Does not react with mouse and rat.

Epitope: aa 1-160

Immunogen: NGFR from A875 melanoma cells

Molecular Weight: 75kDa

Positive Control: Melanoma

Cellular Localization: Cell surface and cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits Binding of NGF to Melanoma Cells

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Neurotrophin Receptor/NGF-Receptor (p75superscriptNGFRsuperscript) Ab-1
Flow Cytometry
Concentrated
RUO
Primary
IgG2a kappa
NGFR5
Monoclonal
Murine
Neuroscience
NGFR from A875 melanoma cells