Please login to your online account to display your discounted pricing

Alfa Aesar™ Neurotensin (9-13)

For cellular and molecular biology applications

Specifications

Product Type Neurotensin (9-13)
Molecular Weight 660.81
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Tyr-Ile-Leu
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66433MA

 
alfa aesar™
J66433MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66433MB

 
alfa aesar™
J66433MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Neurotensin (9-13)
Molecular Weight 660.81
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Tyr-Ile-Leu
Format Solid
Regulatory Status RUO
Purification Method HPLC