Please login to your online account to display your discounted pricing

Alfa Aesar™ Neuropeptide Y (3-36), Human

For cellular and molecular biology applications

Specifications

Product Type Neuropeptide Y (3-36), human
Cross Reactivity Human
Molecular Weight 4011.44
Storage Requirements -30° to -10°C
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66381EXD

 
alfa aesar™
J66381EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66381MCR

 
alfa aesar™
J66381MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Neuropeptide Y (3-36), human
Cross Reactivity Human
Molecular Weight 4011.44
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
Purification Method HPLC