Please login to your online account to display your discounted pricing

Alfa Aesar™ Neuropeptide Y (13-36), Human

For cellular and molecular biology applications

Specifications

Product Type Neuropeptide Y (13-36), human
Cross Reactivity Human
Molecular Weight 3000.43
Storage Requirements -30° to -10°C
Sequence Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66612EXD

 
alfa aesar™
J66612EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66612MCR

 
alfa aesar™
J66612MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Neuropeptide Y (13-36), human
Cross Reactivity Human
Molecular Weight 3000.43
Storage Requirements -30° to -10°C
Sequence Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC