Please login to your online account to display your discounted pricing

Alfa Aesar™ Neuropeptide W-23, Human

For cellular and molecular biology applications

Specifications

Product Type Neuropeptide W-23, human
Cross Reactivity Human
Molecular Weight 2584.04
Storage Requirements -30° to -10°C
Sequence Trp-Tyr-Lys-His-Val-Ala-Ser-Pro-Arg-Tyr-His-Thr-Val-Gly-Arg-Ala-Ala-Gly-Leu-Leu-Met-Gly-Leu
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66112EXD

 
alfa aesar™
J66112EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66112MCR

 
alfa aesar™
J66112MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Neuropeptide W-23, human
Cross Reactivity Human
Molecular Weight 2584.04
Storage Requirements -30° to -10°C
Sequence Trp-Tyr-Lys-His-Val-Ala-Ser-Pro-Arg-Tyr-His-Thr-Val-Gly-Arg-Ala-Ala-Gly-Leu-Leu-Met-Gly-Leu
Format Solid
Regulatory Status RUO
Purification Method HPLC