Please login to your online account to display your discounted pricing

Alfa Aesar™ Neuropeptide S, Mouse

For cellular and molecular biology applications

Specifications

Product Type Neuropeptide S, mouse
Cross Reactivity Mouse
Molecular Weight 2182.47
Storage Requirements -30° to -10°C
Sequence Ser-Phe-Arg-Asn-Gly-Val-Gly-Ser-Gly-Ala-Lys-Lys-Thr-Ser-Phe-Arg-Arg-Ala-Lys-Gln
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66563EXD

 
alfa aesar™
J66563EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66563EXO

 
alfa aesar™
J66563EXO
0.2mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66563MCR

 
alfa aesar™
J66563MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Neuropeptide S, mouse
Cross Reactivity Mouse
Molecular Weight 2182.47
Storage Requirements -30° to -10°C
Sequence Ser-Phe-Arg-Asn-Gly-Val-Gly-Ser-Gly-Ala-Lys-Lys-Thr-Ser-Phe-Arg-Arg-Ala-Lys-Gln
Format Solid
Regulatory Status RUO
Purification Method HPLC