Please login to your online account to display your discounted pricing

Alfa Aesar™ Neuropeptide S, Human

For cellular and molecular biology applications

Specifications

Product Type Neuropeptide S, human
Cross Reactivity Human
Molecular Weight 2187.50
Storage Requirements -30° to -10°C
Sequence Ser-Phe-Arg-Asn-Gly-Val-Gly-Thr-Gly-Met-Lys-Lys-Thr-Ser-Phe-Gln-Arg-Ala-Lys-Ser
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66262EXD

 
alfa aesar™
J66262EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66262EXO

 
alfa aesar™
J66262EXO
0.2mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66262MCR

 
alfa aesar™
J66262MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Neuropeptide S, human
Cross Reactivity Human
Molecular Weight 2187.50
Storage Requirements -30° to -10°C
Sequence Ser-Phe-Arg-Asn-Gly-Val-Gly-Thr-Gly-Met-Lys-Lys-Thr-Ser-Phe-Gln-Arg-Ala-Lys-Ser
Format Solid
Regulatory Status RUO
Purification Method HPLC