Please login to your online account to display your discounted pricing

Alfa Aesar™ Neuromedin U, Rat

For cellular and molecular biology applications

Specifications

Product Type Neuromedin U, rat
Cross Reactivity Rat
Molecular Weight 2634.00
Storage Requirements -30° to -10°C
Sequence Tyr-Lys-Val-Asn-Glu-Tyr-Gln-Gly-Pro-Val-Ala-Pro-Ser-Gly-Gly-Phe-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66682EXD

 
alfa aesar™
J66682EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66682MCR

 
alfa aesar™
J66682MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Neuromedin U, rat
Cross Reactivity Rat
Molecular Weight 2634.00
Storage Requirements -30° to -10°C
Sequence Tyr-Lys-Val-Asn-Glu-Tyr-Gln-Gly-Pro-Val-Ala-Pro-Ser-Gly-Gly-Phe-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC