Please login to your online account to display your discounted pricing

Alfa Aesar™ Neurokinin B


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66607EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66607EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Format Solid
Sequence Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Common Name Neuromedin K
Product Type Neurokinin B
Molecular Weight 1210.43
Purification Method HPLC