Please login to your online account to display your discounted pricing

Alfa Aesar™ Neurokinin B

For cellular and molecular biology applications

$158.54 - $158.54

Specifications

Product Type Neurokinin B
Molecular Weight 1210.43
Storage Requirements -30° to -10°C
Sequence Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66607EXD

 
alfa aesar™
J66607EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66607MCR

 
alfa aesar™
J66607MCR
1mg Each for $158.54
Description & Specifications

Specifications

Product Type Neurokinin B
Molecular Weight 1210.43
Storage Requirements -30° to -10°C
Sequence Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2
Format Solid
Common Name Neuromedin K
Regulatory Status RUO
Purification Method HPLC