Please login to your online account to display your discounted pricing

Alfa Aesar™ Neurokinin A

For cellular and molecular biology applications

$142.69 - $142.69

Specifications

Product Type Neurokinin A
Molecular Weight 1133.33
Storage Requirements -30° to -10°C
Format Powder
Sequence His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66817EXD

 
alfa aesar™
J66817EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66817MCR

 
alfa aesar™
J66817MCR
1mg Each for $142.69
Description & Specifications

Specifications

Product Type Neurokinin A
Molecular Weight 1133.33
Storage Requirements -30° to -10°C
Format Powder
Sequence His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
Common Name Substance K
Regulatory Status RUO
Purification Method HPLC