Please login to your online account to display your discounted pricing

Alfa Aesar™ Neurokinin A


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66817MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66817MCR

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Format Powder
Sequence His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Common Name Substance K
Product Type Neurokinin A
Molecular Weight 1133.33
Purification Method HPLC