Eisco NeuLog™ Accessories
For use with all Neuron NeuLog Logger sensors.
Manufacturer:  Eisco BAT200
 View more versions of this product
Catalog No. S98243-44ND
Description
Description
Battery Module
  • Required to use sensors remotely or in the field
  • Powers sensors or chain of sensors operating off-line; connects sensors to RF Communication Module (S98243-46ND)
  • Rechargeable via PC USB port and data cable (sold separately)
  • Features LED charge indicator and push-button operation
Carrying Case
  • Constructed of black ballistic nylon
  • Holds up to 25 probes including cables and accessories
  • Measures 16 × 12 × 2.5 in. (40.6 × 30.5 × 6.4cm)
  • With shoulder strap
Specifications
Specifications
Battery Module
Supplies power to a sensor, or chain of sensors, operating in the Off-Line mode, and to sensors connected to an RF communication module
NeuLog neuron logger sensors
Documents