Eisco™ NeuLog™ USB Connection Module
The USB module is the first in a chain of sensors connected to a USB port of the PC. It provides both power from the PC to the sensors and communication between the PC and the sensors. It operates with OSX, XP, Vista and Windows7. Connects to a PC using a standard USB - mini USB cable (included).
Supplier: Eisco™ USB200
Alert:
This item is intended for educational purposes only.
Description
- Enables fast connection of Neulog sensors to a PC
- Provides sensors with power and communication to a PC
- Operates with OSX, XP, Vista, and Windows 7
- Standard USB-mini USB connection cable required, included
Specifications
USB Module | |
USB to micro USB cable | |
1 | |
NeuLog neuron logger sensors |
OSX, XP, Vista, Windows7 | |
USB-microUSB | |
Connects the whole NeuLog sensor line and RF module, to any computer with a USB port. |