NeuLog™ Accessories
For use with all Neuron NeuLog Logger sensors.
$63.45 - $74.95
Specifications
For Use With (Equipment) NeuLog neuron logger sensors
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
S98243-44ND Fisher Scientific
BAT-200
Battery Module Each for $74.95
S98243-45ND Fisher Science Education
USB-200
USB Module Each for $63.45
Specifications
Description
Battery Module
  • Required to use sensors remotely or in the field
  • Powers sensors or chain of sensors operating off-line; connects sensors to RF Communication Module (S98243-46ND)
  • Rechargeable via PC USB port and data cable (sold separately)
  • Features LED charge indicator and push-button operation
Carrying Case
  • Constructed of black ballistic nylon
  • Holds up to 25 probes including cables and accessories
  • Measures 16 × 12 × 2.5 in. (40.6 × 30.5 × 6.4cm)
  • With shoulder strap
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
NeuLog neuron logger sensors