Learn More
Netilmicin sulfate, 595 ^mg/mg
A broad spectrum antibiotic | CAS: 56391-57-2 | C21H43N5O11S | 573.659 g/mol
$360.12 - $360.12
Chemical Identifiers
| CAS | 56391-57-2 |
|---|---|
| Molecular Formula | C21H43N5O11S |
| Molecular Weight (g/mol) | 573.659 |
| MDL Number | MFCD01311715 |
| InChI Key | PQHZSWZPVGHDEZ-UIQTUGNFSA-N |
| Synonym | 1-N-Ethylsisomicin |
| PubChem CID | 23581789 |
| IUPAC Name | (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4-amino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-6-(ethylamino)-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid |
| SMILES | CCNC1CC(C(C(C1OC2C(C(C(CO2)(C)O)NC)O)O)OC3C(CC=C(O3)CN)N)N.OS(=O)(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ66302MA
|
Thermo Scientific Chemicals
J66302MA |
10 mg |
Each for $360.12
|
|
|||||
Description
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNetilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied
Solubility
Soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Chemical Identifiers
| 56391-57-2 | |
| 573.659 | |
| PQHZSWZPVGHDEZ-UIQTUGNFSA-N | |
| 23581789 | |
| CCNC1CC(C(C(C1OC2C(C(C(CO2)(C)O)NC)O)O)OC3C(CC=C(O3)CN)N)N.OS(=O)(=O)O |
| C21H43N5O11S | |
| MFCD01311715 | |
| 1-N-Ethylsisomicin | |
| (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4-amino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-6-(ethylamino)-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid |
Specifications
| Netilmicin Sulfate | |
| 56391-57-2 | |
| 10 mg | |
| C21H43N5O11S | |
| 868°C | |
| 1-N-Ethylsisomicin | |
| PQHZSWZPVGHDEZ-UIQTUGNFSA-N | |
| (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4-amino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-6-(ethylamino)-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid | |
| 23581789 | |
| Netilmicin sulfate, 590μg/mg |
| Powder | |
| White | |
| Hygroscopic; Light sensitive | |
| MFCD01311715 | |
| 14,6479 | |
| Soluble in water | |
| CCNC1CC(C(C(C1OC2C(C(C(CO2)(C)O)NC)O)O)OC3C(CC=C(O3)CN)N)N.OS(=O)(=O)O | |
| 573.659 | |
| 720.75 |
Safety and Handling
GHS H Statement
H360-H312-H332
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P281-P308+P313-P501c
H360
EINECSNumber : 260-147-3
RTECSNumber : WK2286000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only