missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nestin Mouse anti-Human, Clone: 10C2, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen Nestin
Clone 10C2
Host Species Murine
Species Reactivity Human
Applications Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Microscopy,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013822 Life Technologies
LS14984380
25μg Each for $158.05
5013823 Life Technologies
LS14984382
100μg Each for $404.55
Specifications

Description

The monoclonal antibody 10C2 recognizes human nestin (residues 1464-1614). Nestin is a 220-240kDa Class VI intermediate filament protein that is expressed in stem cells of the developing nervous system. Expression has also been found in pancreatic islet and mesenchymal precursors in addition to cells with angiogenic potential. Upon differentiation, nestin-containing filaments are replaced by cell type-specific intermediate filament proteins, such as GFAP. Expression has been identified in a variety of cancerous cells including glioblastomas and pancreatic tumors. No cross-reactivity to rodent nestins has been observed using the 10C2 antibody.
Description & Specifications

Specifications

Nestin
Murine
Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Microscopy,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
10C2
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer