Please login to your online account to display your discounted pricing

Nessler Reagent, for Ammonia Nitrogen Determination, Ricca Chemical


Quantity 4L
Packaging Amber Plastic Bottle

Manufacturer: ricca chemical company  RIR52500004B

 View more versions of this product

Catalog No. R52500004B

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Nessler Reagent
CAS 7732-18-5
CAS Min % 73.0
CAS Max % 76.0
CAS 1310-73-2
CAS Min % 14.0
CAS Max % 18.0
CAS 7681-11-0
CAS Min % 3.0
CAS Max % 9.0
CAS 7774-29-0
CAS Min % 4.0
CAS Max % 12.0
Packaging Amber Plastic Bottle
Physical Form Liquid
Color Clear light yellow
Quantity 4L

Specifications

Chemical Name or Material Nessler Reagent
CAS 7732-18-5
CAS Min % 73.0
CAS Max % 76.0
CAS 1310-73-2
CAS Min % 14.0
CAS Max % 18.0
CAS 7681-11-0
CAS Min % 3.0
CAS Max % 9.0
CAS 7774-29-0
CAS Min % 4.0
CAS Max % 12.0
Physical Form Liquid