missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Neryl acetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1928222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Neryl acetate | |
0.91 | |
98°C (208°F) | |
C12H20O2 | |
1.46 | |
100 g | |
neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h | |
CC(=CCCC(=CCOC(=O)C)C)C | |
196.29 | |
196.29 |
141-12-8 | |
133°C to 135°C (25 mmHg) | |
Pleasant | |
(CH3)2C=CHCH2CH2(CH3)C=CHCH2CO2CH3 | |
MFCD00063205 | |
1722814 | |
HIGQPQRQIQDZMP-FLIBITNWSA-N | |
[(2Z)-3,7-dimethylocta-2,6-dienyl] acetate | |
1549025 | |
98% |
Chemical Identifiers
141-12-8 | |
196.29 | |
HIGQPQRQIQDZMP-FLIBITNWSA-N | |
1549025 | |
CC(=CCCC(=CCOC(=O)C)C)C |
C12H20O2 | |
MFCD00063205 | |
neryl acetate, nerol acetate, neryl ethanoate, z-3,7-dimethylocta-2,6-dien-1-yl acetate, nerol acetate 6ci, unii-of82iju18h, cis-3,7-dimethyl-2,6-octadien-1-yl acetate, cis-geranyl acetate, cis-3,7-dimethyl-2,6-octadien-1-ol acetate, of82iju18h | |
[(2Z)-3,7-dimethylocta-2,6-dienyl] acetate |
Safety and Handling
EINECSNumber : 205-459-2
RTECSNumber : RG5921000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only