missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nepsilon-(tert-Butoxycarbonyl)-L-lysine, TCI America™

$94.72 - $273.48

Chemical Identifiers

CAS 2418-95-3
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.307
MDL Number MFCD00037221
InChI Key VVQIIIAZJXTLRE-QMMMGPOBSA-N
Synonym h-lys boc-oh, lys boc, nepsilon-tert-butoxycarbonyl-l-lysine, n6-boc-l-lysine, n~6~-tert-butoxycarbonyl-l-lysine, nepsilon-boc-l-lysine, ne-boc-l-lysine, n epsilon-boc-l-lysine, h-lys boc-2-chlorotrityl resin, n6-tert-butoxycarbonyl-l-lysine
PubChem CID 2733283
IUPAC Name (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCCC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16705G View Documents TCI America
B16705G
5g Each for $273.48
 
B16701G View Documents TCI America
B16701G
1g Each for $94.72
 
Specifications

Chemical Identifiers

2418-95-3
246.307
VVQIIIAZJXTLRE-QMMMGPOBSA-N
2733283
CC(C)(C)OC(=O)NCCCCC(C(=O)O)N
C11H22N2O4
MFCD00037221
h-lys boc-oh, lys boc, nepsilon-tert-butoxycarbonyl-l-lysine, n6-boc-l-lysine, n~6~-tert-butoxycarbonyl-l-lysine, nepsilon-boc-l-lysine, ne-boc-l-lysine, n epsilon-boc-l-lysine, h-lys boc-2-chlorotrityl resin, n6-tert-butoxycarbonyl-l-lysine
(2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid

Specifications

2418-95-3
MFCD00037221
VVQIIIAZJXTLRE-QMMMGPOBSA-N
(2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
2733283
Crystal-Powder at 20°C
White
Nepsilon-(tert-Butoxycarbonyl)-L-lysine
C11H22N2O4
h-lys boc-oh, lys boc, nepsilon-tert-butoxycarbonyl-l-lysine, n6-boc-l-lysine, n~6~-tert-butoxycarbonyl-l-lysine, nepsilon-boc-l-lysine, ne-boc-l-lysine, n epsilon-boc-l-lysine, h-lys boc-2-chlorotrityl resin, n6-tert-butoxycarbonyl-l-lysine
CC(C)(C)OC(=O)NCCCCC(C(=O)O)N
246.307
246.31
≥97.0% (T)
5g
Safety and Handling

Safety and Handling

TSCA : No