missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Fmoc-L-lysine, 98%

$28.07 - $326.93

Chemical Identifiers

CAS 84624-28-2
Molecular Formula C21H24N2O4
Molecular Weight (g/mol) 368.433
MDL Number MFCD00038365
InChI Key RAQBUPMYCNRBCQ-IBGZPJMESA-N
Synonym h-lys fmoc-oh, n'-fmoc-l-lysine, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-aminohexanoic acid, n6-fmoc-l-lysine, lys fmoc-oh, n-, a-fmoc-l-lysine, n-epsilon-9-fluorenylmethyloxycarbonyl-l-lysine, l-lysine, n6-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
PubChem CID 7010541
IUPAC Name (2S)-2-amino-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62834MD View Documents Alfa Aesar™
H62834MD
250mg Each for $28.07
 
AAH6283403 View Documents Alfa Aesar™
H6283403
1g Each for $82.50
 
AAH6283406 View Documents Alfa Aesar™
H6283406
5g Each for $326.93
 
Specifications

Chemical Identifiers

84624-28-2
368.433
RAQBUPMYCNRBCQ-IBGZPJMESA-N
7010541
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)N
C21H24N2O4
MFCD00038365
h-lys fmoc-oh, n'-fmoc-l-lysine, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-aminohexanoic acid, n6-fmoc-l-lysine, lys fmoc-oh, n-, a-fmoc-l-lysine, n-epsilon-9-fluorenylmethyloxycarbonyl-l-lysine, l-lysine, n6-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
(2S)-2-amino-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid

Specifications

250mg
C21H24N2O4
h-lys fmoc-oh, n'-fmoc-l-lysine, s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-aminohexanoic acid, n6-fmoc-l-lysine, lys fmoc-oh, n-, a-fmoc-l-lysine, n-epsilon-9-fluorenylmethyloxycarbonyl-l-lysine, l-lysine, n6-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCC(C(=O)O)N
368.433
368.43
Nepsilon-Fmoc-L-lysine
84624-28-2
MFCD00038365
RAQBUPMYCNRBCQ-IBGZPJMESA-N
(2S)-2-amino-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
7010541
98%