missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Boc-Nalpha-Fmoc-Nepsilon-isopropyl-L-lysine, 94%

$252.91 - $760.27

Chemical Identifiers

CAS 201003-48-7
Molecular Formula C29H38N2O6
Molecular Weight (g/mol) 510.631
MDL Number MFCD00080276
InChI Key LUGFCMICCJNLBC-VWLOTQADSA-N
Synonym n-fmoc-n'-boc-n'-isopropyl-l-lysine, fmoc-lys boc isopropyl-oh, fmoc-lys ipr,boc-oh, 2s-6-tert-butoxycarbonyl isopropyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1447, fmoc-l-lys boc, ipr-oh, n-9-fluorenylmethyloxycarbonyl-n'-tert-butoxycarbonyl-n'-isopropyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl isopropyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-n-epsilon-i-propyl-l-lysine, n~6~-tert-butoxycarbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-propan-2-yl-l-lysine
PubChem CID 51340513
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonyl-propan-2-ylamino]hexanoic acid
SMILES CC(C)N(CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66313MD View Documents Alfa Aesar™
H66313MD
250mg Each for $252.91
 
AAH6631303 View Documents Alfa Aesar™
H6631303
1g Each for $760.27
 
Specifications

Chemical Identifiers

201003-48-7
510.631
LUGFCMICCJNLBC-VWLOTQADSA-N
51340513
CC(C)N(CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)OC(C)(C)C
C29H38N2O6
MFCD00080276
n-fmoc-n'-boc-n'-isopropyl-l-lysine, fmoc-lys boc isopropyl-oh, fmoc-lys ipr,boc-oh, 2s-6-tert-butoxycarbonyl isopropyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1447, fmoc-l-lys boc, ipr-oh, n-9-fluorenylmethyloxycarbonyl-n'-tert-butoxycarbonyl-n'-isopropyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl isopropyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-n-epsilon-i-propyl-l-lysine, n~6~-tert-butoxycarbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-propan-2-yl-l-lysine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonyl-propan-2-ylamino]hexanoic acid

Specifications

201003-48-7
MFCD00080276
LUGFCMICCJNLBC-VWLOTQADSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonyl-propan-2-ylamino]hexanoic acid
51340513
Solid
250mg
C29H38N2O6
n-fmoc-n'-boc-n'-isopropyl-l-lysine, fmoc-lys boc isopropyl-oh, fmoc-lys ipr,boc-oh, 2s-6-tert-butoxycarbonyl isopropyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1447, fmoc-l-lys boc, ipr-oh, n-9-fluorenylmethyloxycarbonyl-n'-tert-butoxycarbonyl-n'-isopropyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl isopropyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-n-epsilon-i-propyl-l-lysine, n~6~-tert-butoxycarbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-n~6~-propan-2-yl-l-lysine
CC(C)N(CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)OC(C)(C)C
510.631
510.63
94%
Nepsilon-Boc-Nalpha-Fmoc-Nepsilon-isopropyl-L-lysine
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C