missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H62993MD

 View more versions of this product

Catalog No. AAH62993MD


 

Description

Description

Applications
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride
95%
MFCD00038899
TZBPQINFXPIRBX-MERQFXBCSA-N
tert-butyl (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate;hydrochloride
13819885
Powder
250mg
13288-57-8
C15H31ClN2O4
H-Lys(Boc)-OtBu.HCl; (S)-tert-Butyl 2-amino-6-(Boc-amino)hexanoate hydrochloride
CC(C)(C)OC(=O)C(CCCCNC(=O)OC(C)(C)C)N.Cl
338.873
338.87
White

Chemical Identifiers

13288-57-8
338.873
TZBPQINFXPIRBX-MERQFXBCSA-N
13819885
C15H31ClN2O4
MFCD00038899
H-Lys(Boc)-OtBu.HCl; (S)-tert-Butyl 2-amino-6-(Boc-amino)hexanoate hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents