Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Fmoc-D-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Fmoc-D-lysine
CAS 110990-07-3
Formula Weight 502.57
MDL Number MFCD00065663
Synonym Fmoc-D-Lys(Z)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6617803

 
alfa aesar™
H6617803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6617806

 
alfa aesar™
H6617806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66178MD

 
alfa aesar™
H66178MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 502.57
Melting Point 106° to 110°C
CAS 110990-07-3
Physical Form Powder
Assay 95%
MDL Number MFCD00065663
Synonym Fmoc-D-Lys(Z)-OH
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Fmoc-D-lysine
Color White
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C