Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%

Manufacturer:  Alfa Aesar™ H6347003

 View more versions of this product

Catalog No. AAH6347003



Description

Description

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester
C23H31N3O8
1559393
YWLICOCXPNQJPC-KRWDZBQOSA-N
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoate
12686578
Powder
White
1g
34404-36-9
MFCD00037915
s-5-11-dimethylethoxy carbonyl amino-6-25-dioxo-1-pyrrolidinyl oxy-6-oxohexyl carbamic acid phenylmethyl ester, a-z-l-lysine hydroxysuccinimide ester, 25-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, s-25-dioxopyrrolidin-1-yl 6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, 25-dioxopyrrolidin-1-yl s-6-benzyloxycarbonyl amino-2-tert-butoxycarbonyl-amino hexanoate, 25-dioxopyrrolidin-1-yl s-6-benzyloxycarbonyl amino-2-tertbutoxycarbonyl amino hexanoate, 25-dioxopyrrolidin-1-yl s-6-benzyloxycarbonyl amino-2-tert-butoxycarbonyl amino hexanoate, 25-dioxopyrrolidin-1-yl n6-benzyloxy carbonyl-n2-tert-butoxycarbonyl-l-lysinate, n, a-boc-n, a-cbz-l-lysine n-hydroxysuccinimide ester
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
477.514
477.51
95%
108°C to 117°C

Chemical Identifiers

34404-36-9
477.514
YWLICOCXPNQJPC-KRWDZBQOSA-N
12686578
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
C23H31N3O8
MFCD00037915
s-5-11-dimethylethoxy carbonyl amino-6-25-dioxo-1-pyrrolidinyl oxy-6-oxohexyl carbamic acid phenylmethyl ester, a-z-l-lysine hydroxysuccinimide ester, 25-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, s-25-dioxopyrrolidin-1-yl 6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, 25-dioxopyrrolidin-1-yl s-6-benzyloxycarbonyl amino-2-tert-butoxycarbonyl-amino hexanoate, 25-dioxopyrrolidin-1-yl s-6-benzyloxycarbonyl amino-2-tertbutoxycarbonyl amino hexanoate, 25-dioxopyrrolidin-1-yl s-6-benzyloxycarbonyl amino-2-tert-butoxycarbonyl amino hexanoate, 25-dioxopyrrolidin-1-yl n6-benzyloxy carbonyl-n2-tert-butoxycarbonyl-l-lysinate, n, a-boc-n, a-cbz-l-lysine n-hydroxysuccinimide ester
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoate
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents