Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%

Chemical Identifiers

Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester
CAS 34404-36-9
Formula Weight 477.51
MDL Number MFCD00037915
Beilstein 1559393
Synonym Boc-Lys(Z)-OSu
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6347003

  SDS
alfa aesar™
H6347003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6347006

  SDS
alfa aesar™
H6347006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6347014

  SDS
alfa aesar™
H6347014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 108° to 117°C
Formula Weight 477.51
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester
Color White
CAS 34404-36-9
Beilstein 1559393
Physical Form Powder
Quantity 1g
Assay 95%
MDL Number MFCD00037915
Storage Note 1 -30° to -10°C
Synonym Boc-Lys(Z)-OSu

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

-30° to -10°C