Learn More
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6347014
Description
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester is a reactant involved in Cellular labeling and tumor MRI as a bimodal lipidic contrast agent, Synthesis of improved peptide prodrugs 5-ALA for photodynamic therapy, Design of high affinity peptide conjugates as markers for small peptide transporter PEPT1 (SLC15A1), Preparation of human growth hormone derivatives containing PEG groups, Enantiodivergent asymmetric cyclization and Solid-phase synthesis of peptide-cationic steroid conjugates for antibacterial screening.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySlightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
108°C to 117°C | |
34404-36-9 | |
C23H31N3O8 | |
1559393 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O | |
477.51 | |
477.51 | |
25 g |
Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine N-succinimidyl ester | |
White | |
MFCD00037915 | |
boc-lys z-osu, a-boc-l-lysine hydroxysuccinimide ester, a-cbz-l-lysine n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys cbz-osu, 2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys-z-osu, boc-lys z-o su, boc-lys-z-osu, n,a-boc-n, a-z-l-lysine hydroxysuccinimide ester, n,a-z-n | |
YWLICOCXPNQJPC-AGDOHHJYNA-N | |
2,5-dioxopyrrolidin-1-yl 6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoate | |
12686578 | |
95% | |
Powder |
Chemical Identifiers
34404-36-9 | |
477.51 | |
YWLICOCXPNQJPC-AGDOHHJYNA-N | |
12686578 | |
CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O |
C23H31N3O8 | |
MFCD00037915 | |
boc-lys z-osu, a-boc-l-lysine hydroxysuccinimide ester, a-cbz-l-lysine n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys cbz-osu, 2,5-dioxopyrrolidin-1-yl 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoate, boc-lys-z-osu, boc-lys z-o su, boc-lys-z-osu, n,a-boc-n, a-z-l-lysine hydroxysuccinimide ester, n,a-z-n | |
2,5-dioxopyrrolidin-1-yl 6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoate |
Safety and Handling
EINECSNumber : 251-999-7
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only