Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride
CAS 6366-70-7
Formula Weight 406.9
MDL Number MFCD00038981
Beilstein 3580519
Synonym H-Lys(Z)-Obzl.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6371403

  SDS
alfa aesar™
H6371403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6371406

  SDS
alfa aesar™
H6371406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 406.9
Assay 98%
Quantity 1g
RTECS Number OL5685000
MDL Number MFCD00038981
Storage Note 1 2° to 8°C
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride
Beilstein 3580519
Synonym H-Lys(Z)-Obzl.HCl
Color White to off-white
Physical Form Powder
CAS 6366-70-7
Safety and Handling

2° to 8°C