Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-L-lysinamide hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material Nepsilon-Benzyloxycarbonyl-L-lysinamide hydrochloride
CAS 58117-53-6
Formula Weight 315.8
MDL Number MFCD00035520
Beilstein 6088865
Synonym H-Lys(Z)-NH2.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6519903

 
alfa aesar™
H6519903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6519906

 
alfa aesar™
H6519906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65199MD

 
alfa aesar™
H65199MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 315.8
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-L-lysinamide hydrochloride
MDL Number MFCD00035520
Assay 95%
Physical Form Solid
Synonym H-Lys(Z)-NH2.HCl
Storage Note 1 2° to 8°C
Beilstein 6088865
Quantity 1g
CAS 58117-53-6
Safety and Handling

2° to 8°C