Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-D-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nepsilon-Benzyloxycarbonyl-D-lysine
CAS 34404-32-5
Formula Weight 280.32
MDL Number MFCD00063120
Synonym H-D-Lys(Z)-OH; Nε-Cbz-L-Lysine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6281903

  SDS
alfa aesar™
H6281903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6281914

  SDS
alfa aesar™
H6281914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6281906

  SDS
alfa aesar™
H6281906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.32
Physical Form Powder
Assay 95%
Color White
Quantity 1g
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-D-lysine
MDL Number MFCD00063120
CAS 34404-32-5
Synonym H-D-Lys(Z)-OH; Nε-Cbz-L-Lysine