missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Benzyloxycarbonyl-D-lysine, 95%

Manufacturer:  Alfa Aesar™ H6281906

 View more versions of this product

Catalog No. AAH6281906


 

Specifications

Specifications

Nepsilon-Benzyloxycarbonyl-D-lysine
95%
MFCD00063120
CKGCFBNYQJDIGS-GFCCVEGCSA-N
(2R)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid
1551330
Powder
5g
34404-32-5
C14H20N2O4
H-D-Lys(Z)-OH; Nε-Cbz-L-Lysine
C1=CC=C(C=C1)COC(=O)NCCCCC(C(=O)O)N
280.324
280.32
White

Chemical Identifiers

34404-32-5
280.324
CKGCFBNYQJDIGS-GFCCVEGCSA-N
1551330
C14H20N2O4
MFCD00063120
H-D-Lys(Z)-OH; Nε-Cbz-L-Lysine
SDS Documents