Please login to your online account to display your discounted pricing

Nepsilon-Benzoyl-L-lysine, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  333470250

 View more versions of this product

Catalog No. AC333470250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 250.3
Quantity 25g
Color White to beige
CAS Max % 100.0
Molecular Formula C13H18N2O3
Infrared Spectrum Authentic
Chemical Name or Material Nε-Benzoyl-L-lysine
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00037222
CAS 1219-46-1
Packaging Glass bottle
CAS Min % 97.5
Specific Rotation +17° to +21° (20°C, 589nm) (c=3.2, 1N HCl)

Specifications

Chemical Name or Material Nε-Benzoyl-L-lysine
Name Note 98%
CAS 1219-46-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H18N2O3
Formula Weight 250.3
MDL Number MFCD00037222