Please login to your online account to display your discounted pricing

Nepsilon-Benzoyl-L-lysine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Nε-Benzoyl-L-lysine
Name Note 98%
CAS 1219-46-1
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C13H18N2O3
Formula Weight 250.3g/mol
MDL Number MFCD00037222
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC333470250

  SDS
thermo fisher scientific
333470250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +17° to +21° (20°C, 589nm) (c=3.2, 1N HCl)
Color White to beige
Quantity 25g
Chemical Name or Material Nε-Benzoyl-L-lysine
Name Note 98%
CAS 1219-46-1
CAS Min % 97.5
CAS Max % 100.0
Physical Form White to beige powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C13H18N2O3
Formula Weight 250.3g/mol
MDL Number MFCD00037222