missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-L-lysine, 95%

$59.51 - $226.69

Chemical Identifiers

CAS 146982-27-6
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.507
MDL Number MFCD00190872
InChI Key OJBNDXHENJDCBA-QFIPXVFZSA-N
Synonym fmoc-lys alloc-oh, a-fmoc-n, a-alloc-l-lysine, fmoc-lys aloc-oh, n-fmoc-n'-allyoxycarbonyl-l-lysine, s-2-9h-ffuoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, fmoc-lys alloc, aloc-lys fmoc-oh, pubchem13153, fmoc-l-lys alloc-oh, n
PubChem CID 11123257
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid
SMILES C=CCOC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6376803 View Documents Alfa Aesar™
H6376803
1g Each for $59.51
 
AAH6376806 View Documents Alfa Aesar™
H6376806
5g Each for $226.69
 
Specifications

Chemical Identifiers

146982-27-6
452.507
OJBNDXHENJDCBA-QFIPXVFZSA-N
11123257
C=CCOC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C25H28N2O6
MFCD00190872
fmoc-lys alloc-oh, a-fmoc-n, a-alloc-l-lysine, fmoc-lys aloc-oh, n-fmoc-n'-allyoxycarbonyl-l-lysine, s-2-9h-ffuoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, fmoc-lys alloc, aloc-lys fmoc-oh, pubchem13153, fmoc-l-lys alloc-oh, n
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid

Specifications

146982-27-6
MFCD00190872
fmoc-lys alloc-oh, a-fmoc-n, a-alloc-l-lysine, fmoc-lys aloc-oh, n-fmoc-n'-allyoxycarbonyl-l-lysine, s-2-9h-ffuoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, fmoc-lys alloc, aloc-lys fmoc-oh, pubchem13153, fmoc-l-lys alloc-oh, n
C=CCOC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
452.507
452.5
95%
Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-L-lysine
C25H28N2O6
5893081
OJBNDXHENJDCBA-QFIPXVFZSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid
11123257
Solid
1g
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C