Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine
CAS 214750-75-1
Formula Weight 452.51
MDL Number MFCD00798639
Synonym Fmoc-D-Lys(Aloc)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6686103

  SDS
alfa aesar™
H6686103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6686106

  SDS
alfa aesar™
H6686106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 452.51
CAS 214750-75-1
MDL Number MFCD00798639
Assay 95%
Chemical Name or Material Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-D-lysine
Synonym Fmoc-D-Lys(Aloc)-OH
Storage Note 1 2° to 8°C
Quantity 1g
Color White
Physical Form Powder
Safety and Handling

2° to 8°C