Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-(2-Chlorobenzyloxycarbonyl)-Nalpha-Fmoc-L-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nepsilon-(2-Chlorobenzyloxycarbonyl)-Nalpha-Fmoc-L-lysine
CAS 133970-31-7
Formula Weight 537.01
MDL Number MFCD00065664
Synonym Fmoc-Lys(2-Cl-Z)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6661506

 
alfa aesar™
H6661506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6661514

 
alfa aesar™
H6661514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 537.01
Chemical Name or Material Nepsilon-(2-Chlorobenzyloxycarbonyl)-Nalpha-Fmoc-L-lysine
MDL Number MFCD00065664
CAS 133970-31-7
Assay 95%
Synonym Fmoc-Lys(2-Cl-Z)-OH
Physical Form Solid
Quantity 5g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C