Please login to your online account to display your discounted pricing

Alfa Aesar™ Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine, 98%

Chemical Identifiers

Chemical Name or Material Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine
CAS 148083-64-1
Formula Weight 534.52
MDL Number MFCD00273454
Synonym Fmoc-Lys(Dnp)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6305003

 
alfa aesar™
H6305003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6305006

 
alfa aesar™
H6305006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63050MD

 
alfa aesar™
H63050MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 534.52
Chemical Name or Material Nepsilon-2,4-Dinitrophenyl-Nalpha-Fmoc-L-lysine
Physical Form Solid
Assay 98%
MDL Number MFCD00273454
CAS 148083-64-1
Synonym Fmoc-Lys(Dnp)-OH
Quantity 1g