Please login to your online account to display your discounted pricing

Neotek™ Virtual Reality System Software


Detailed information in fun 3-D format.

Manufacturer: fisher science education  2004

 View more versions of this product

Catalog No. S66196

  • / Each
 

Description & Specifications

Specifications

Compatibility Windows™ 95, 98, ME, NT, 2000, XP™
For Use With (Application) Intended for the anatomy and physiology student that wants the same level of understanding achieved in the lab
Format CD-ROM
Grade College
Includes 30 gross anatomy dissections
Memory 2MB
Classification Life science
Product Type The Cell is A City Software