Please login to your online account to display your discounted pricing

Neotek™ Virtual Reality System Software


Detailed information in fun 3-D format.

Manufacturer: fisher science education  2101EMD

 View more versions of this product

Catalog No. S66205

  • / Each
 

Description & Specifications

Specifications

Includes Materials from NASA
Classification Space science
Product Type Mission To Mars Software
Compatibility Windows™ 95, 98, ME, NT, 2000, XP™
For Use With (Application) Provide the ultimate teaching and learning experience by combining 3-D technology with the seriousness of in-depth
Format CD-ROM
Memory 2MB