Please login to your online account to display your discounted pricing

Alfa Aesar™ Neopentylbenzene, 97+%

Chemical Identifiers

Chemical Name or Material Neopentylbenzene
CAS 1007-26-7
Molecular Formula C11H16
Formula Weight 148.25
MDL Number MFCD00038279
UN Number UN3295
Beilstein 2037544
Synonym 2,2-Dimethyl-1-phenylpropane; 2,2-Dimethylpropylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1294114

  SDS
alfa aesar™
A1294114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.25
Chemical Name or Material Neopentylbenzene
Assay ≥97%
Molecular Formula C11H16
MDL Number MFCD00038279
CAS 1007-26-7
UN Number UN3295
Boiling Point 184° to 185°C
Beilstein 2037544
Quantity 25g
Flash Point 50°C (122°F)
Synonym 2,2-Dimethyl-1-phenylpropane; 2,2-Dimethylpropylbenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

Applications
It is used in the preparation of α-Disulfones by Cobalt(III) Oxidation.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.