Please login to your online account to display your discounted pricing

Neomycin Sulfate (White Powder), Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP26695

 View more versions of this product

Catalog No. BP26695

  • / Each
  • / Case of 18 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 614.65
Quantity 5g
Physical Form Solid
Molecular Formula C23H46N6O13 · 3 H2SO4
Color White
pH 6.8
Merck Index 15, 6539
Loss on Drying < 8%
Assay Percent Range > 98.5 %
Health Hazard 2 WARNING!
ChemAlert Storage Symbol Gray
CAS 1405-10-3
Packaging Amber Glass
Synonym Neobiotic
Purity > 98.5%
Identification Pass Test
Potency > 650μg/mg
Health Hazard 3 Emergency Overview
Substances known to cause developmental toxicity in humans. May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:1
Recommended Storage RT, desiccate, protect from light.
Chemical Name or Material Neomycin Sulfate

Specifications

Chemical Name or Material Neomycin Sulfate
CAS 1405-10-3
Physical Form Solid
Assay Percent Range > 98.5 %
Molecular Formula C23H46N6O13 · 3 H2SO4
Formula Weight 614.65
Synonym Neobiotic
Accessories
 

Emergency Overview
Substances known to cause developmental toxicity in humans. May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:1

WARNING!

RT, desiccate, protect from light.

SDS & Safety Documents